siRNA Details
| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178160.2 | NP_835454.1 | 6 | 1137 | ||
| XM_011525479.1 | XP_011523781.1 | 7 | 1347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTOP2| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178160.2 | NP_835454.1 | 6 | 1137 | ||
| XM_011525479.1 | XP_011523781.1 | 7 | 1347 | |||