siRNA Details
| Gene Symbol: SLC38A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033518.3 | NP_277053.2 | 11 | 866 | ||
| XM_005272694.3 | XP_005272751.2 | 10 | 856 | |||
| XM_005272695.4 | XP_005272752.3 | 10 | 858 | |||
| XM_005272697.2 | XP_005272754.2 | 10 | 809 | |||
| XM_005272698.4 | XP_005272755.2 | 10 | 747 | |||
| XM_006724569.3 | XP_006724632.1 | 11 | 866 | |||
| XM_017029960.1 | XP_016885449.1 | 10 | 778 | |||
| XM_017029961.1 | XP_016885450.1 | 10 | 750 | |||
| GenBank mRNA | AF276889.1 | - | 10 | 1531 | ||
| AF318358.1 | - | 7 | 977 | |||
| AK094788.1 | - | 10 | 785 | |||
| BC019246.2 | - | 10 | 716 | 3506044 | ||
| BC027721.2 | - | 6 | 407 | |||

SLC38A5
Genomic Map