siRNA Details
| Gene Symbol: DIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000792.6 | NP_000783.2 | 4 | 1285 | ||
| NM_001039715.2 | NP_001034804.1 | 3 | 1141 | |||
| NM_001039716.2 | NP_001034805.1 | 3 | 1085 | |||
| NM_001324316.1 | NP_001311245.1 | 2 | 941 | |||
| NM_213593.4 | NP_998758.1 | 4 | 1093 | |||
| NR_136692.1 | 4 | 1176 | ||||
| NR_136693.1 | 5 | 1202 | ||||
| GenBank mRNA | BC017955.1 | - | 3 | 1084 | 4607072 | |

DIO1
Genomic Map