siRNA Details
| Gene Symbol: HELB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033647.4 | NP_387467.2 | 2 | 324 | ||
| NR_135080.1 | 2 | 324 | ||||
| NR_135081.1 | 2 | 324 | ||||
| XM_005269234.2 | XP_005269291.1 | 2 | 302 | |||
| XM_017020230.1 | XP_016875719.1 | 2 | 302 | |||
| GenBank mRNA | AA070301.1 | - | 2 | 296 | ||
| AF319995.1 | - | 2 | 265 | |||
| AK290894.1 | - | 2 | 289 | |||
| BC143409.1 | - | 2 | 290 | |||
| BC143411.1 | - | 2 | 290 | |||
| DN997712.1 | - | 2 | 333 | |||

HELB
Genomic Map