siRNA Details
| Gene Symbol: CENPBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145039.3 | NP_659476.2 | 1 | 974 | ||
| GenBank mRNA | AK056131.1 | - | 1 | 612 | ||
| AK074351.1 | - | 1 | 686 | |||
| AK315352.1 | - | 1 | 607 | |||
| CD518450.1 | - | 1 | 12 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPBD1| Gene Symbol: CENPBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145039.3 | NP_659476.2 | 1 | 974 | ||
| GenBank mRNA | AK056131.1 | - | 1 | 612 | ||
| AK074351.1 | - | 1 | 686 | |||
| AK315352.1 | - | 1 | 607 | |||
| CD518450.1 | - | 1 | 12 | |||