siRNA Details
| Gene Symbol: REEP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138393.1 | NP_612402.1 | 2, 3 | 310 | ||
| GenBank mRNA | AK058112.1 | - | 3, 4 | 285 | ||
| AK293919.1 | - | 2, 3 | 283 | |||
| AK315744.1 | - | 2, 3 | 285 | |||
| AX775999.1 | - | 2, 3 | 310 | |||
| AY562244.1 | - | 2, 3 | 206 | |||
| BC008201.1 | - | 2, 3 | 310 | 3865853 | ||

REEP6
Genomic Map