siRNA Details
| Gene Symbol: CCDC102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033212.3 | NP_149989.2 | 9 | 1784 | ||
| XM_011523469.2 | XP_011521771.1 | 9 | 3064 | |||
| GenBank mRNA | BC004307.1 | - | 6 | 741 | ||
| BC008285.1 | - | 9 | 1763 | 3637387 | ||
| BC009941.2 | - | 9 | 1784 | 4100726 | ||

CCDC102A
Genomic Map