siRNA Details
| Gene Symbol: ZBTB47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145166.3 | NP_660149.2 | 2 | 1611 | ||
| GenBank mRNA | AK127065.1 | - | 3 | 1305 | ||
| AL133062.1 | - | 1 | 304 | |||
| BC021855.1 | - | 1 | 269 | 4943271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB47| Gene Symbol: ZBTB47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145166.3 | NP_660149.2 | 2 | 1611 | ||
| GenBank mRNA | AK127065.1 | - | 3 | 1305 | ||
| AL133062.1 | - | 1 | 304 | |||
| BC021855.1 | - | 1 | 269 | 4943271 | ||