siRNA Details
| Gene Symbol: ORAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152288.2 | NP_689501.1 | 2 | 598 | ||
| GenBank mRNA | AK298276.1 | - | 2 | 560 | ||
| BC006126.1 | - | 2 | 541 | 3603998 | ||
| BC015555.1 | - | 2 | 587 | 3878011 | ||
| BC022786.1 | - | 2 | 541 | 3605904 | ||
| BI561433.1 | - | 2 | 621 | |||

ORAI3
Genomic Map