siRNA Details
| Gene Symbol: DIO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001362.3 | NP_001353.4 | 1 | 1778 | ||
| GenBank mRNA | BC017717.1 | - | 1 | 1626 | 4395664 | |
| BX161470.1 | - | 1 | 1778 | |||
| S79854.1 | - | 2 | 1734 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIO3| Gene Symbol: DIO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001362.3 | NP_001353.4 | 1 | 1778 | ||
| GenBank mRNA | BC017717.1 | - | 1 | 1626 | 4395664 | |
| BX161470.1 | - | 1 | 1778 | |||
| S79854.1 | - | 2 | 1734 | |||