siRNA Details
| Gene Symbol: C1orf158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152290.3 | NP_689503.3 | 1 | 334 | ||
| XM_011542434.1 | XP_011540736.1 | 1 | 319 | |||
| GenBank mRNA | AK298760.1 | - | 1 | 296 | ||
| BC029894.1 | - | 1 | 305 | 5171302 | ||
| HY002971.1 | - | 1 | 335 | |||

C1orf158
Genomic Map