siRNA Details
| Gene Symbol: ARMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199196.1 | NP_001186125.1 | 5 | 1052 | ||
| NM_033415.3 | NP_219483.1 | 4 | 944 | |||
| XM_005260157.3 | XP_005260214.1 | 4 | 995 | |||
| XM_017027484.1 | XP_016882973.1 | 4 | 919 | |||
| XM_017027485.1 | XP_016882974.1 | 6 | 1176 | |||
| GenBank mRNA | AK295488.1 | - | 4 | 718 | ||
| AK303914.1 | - | 2 | 632 | |||
| BC056161.1 | - | 6 | 674 | |||
| BC067125.1 | - | 5 | 821 | 6161112 | ||
| BX648486.1 | - | 4 | 839 | |||
| CR749678.1 | - | 1 | 285 | |||

ARMC6
Genomic Map