siRNA Details
| Gene Symbol: ARMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199196.1 | NP_001186125.1 | 5 | 1244 | ||
| NM_033415.3 | NP_219483.1 | 4 | 1136 | |||
| XM_005260157.3 | XP_005260214.1 | 4 | 1187 | |||
| XM_017027484.1 | XP_016882973.1 | 4 | 1111 | |||
| XM_017027485.1 | XP_016882974.1 | 6 | 1368 | |||
| GenBank mRNA | AK295488.1 | - | 4 | 910 | ||
| AK303914.1 | - | 2 | 824 | |||
| BC056161.1 | - | 6 | 866 | |||
| BC067125.1 | - | 5 | 1013 | 6161112 | ||
| BX648486.1 | - | 4 | 1031 | |||
| CR749678.1 | - | 1 | 477 | |||

ARMC6
Genomic Map