siRNA Details
| Gene Symbol: ZNF670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204220.1 | NP_001191149.1 | 4 | 667 | ||
| NM_033213.4 | NP_149990.1 | 4 | 670 | |||
| GenBank mRNA | AK074774.1 | - | 4 | 617 | ||
| BC005360.1 | - | 4 | 617 | 3684736 | ||

ZNF670
Genomic Map