siRNA Details
| Gene Symbol: TBC1D31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145088.1 | NP_001138560.1 | 14 | 1998 | ||
| NM_145647.3 | NP_663622.2 | 14 | 1998 | |||
| XM_005251102.1 | XP_005251159.1 | 14 | 1998 | |||
| XM_005251103.1 | XP_005251160.1 | 14 | 1998 | |||
| XM_011517378.2 | XP_011515680.1 | 14 | 2513 | |||
| XM_011517379.2 | XP_011515681.1 | 14 | 1998 | |||
| XM_017013983.1 | XP_016869472.1 | 14 | 1998 | |||
| XM_017013984.1 | XP_016869473.1 | 13 | 2120 | |||
| XM_017013985.1 | XP_016869474.1 | 13 | 1993 | |||
| XM_017013986.1 | XP_016869475.1 | 13 | 1854 | |||
| XM_017013988.1 | XP_016869477.1 | 13 | 1833 | |||
| XM_017013989.1 | XP_016869478.1 | 13 | 1996 | |||
| XM_017013990.1 | XP_016869479.1 | 12 | 1662 | |||
| GenBank mRNA | AK056434.1 | - | 14 | 1999 | ||
| AK094612.1 | - | 13 | 1796 | |||
| BC027237.1 | - | 3 | 270 | |||
| BC101724.1 | - | 14 | 1965 | 8069230 | ||
| BC111954.1 | - | 14 | 1965 | 8327422 | ||
| BC143523.1 | - | 13 | 1849 | |||
| BC143525.1 | - | 14 | 1965 | 9052034 | ||

TBC1D31
Genomic Map