siRNA Details
| Gene Symbol: ARHGAP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033515.2 | NP_277050.2 | 12 | 1762 | ||
| XM_005267212.2 | XP_005267269.2 | 12 | 1838 | |||
| XM_005267213.1 | XP_005267270.1 | 12 | 1762 | |||
| XM_017011494.1 | XP_016866983.1 | 12 | 1838 | |||
| GenBank mRNA | AB053293.1 | - | 11 | 1560 | ||
| AL834511.1 | - | 12 | 1700 | |||
| BC101708.1 | - | 12 | 1756 | 8069214 | ||
| BC107416.1 | - | 12 | 1718 | 4040931 | ||
| BC111940.1 | - | 12 | 1756 | 8327408 | ||
| BC143531.1 | - | 12 | 1756 | |||

ARHGAP18
Genomic Map