siRNA Details
| Gene Symbol: ACRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052957.4 | NP_443189.1 | 12 | 2309 | ||
| XM_006724716.3 | XP_006724779.1 | 12 | 1862 | |||
| XM_011531063.2 | XP_011529365.1 | 11 | 2129 | |||
| XM_011531064.1 | XP_011529366.1 | 12 | 1912 | |||
| XM_011531066.2 | XP_011529368.1 | 9 | 2048 | |||
| XM_011531067.2 | XP_011529369.1 | 9 | 2048 | |||
| XM_017029965.1 | XP_016885454.1 | 13 | 1963 | |||
| GenBank mRNA | AJ311392.1 | - | 11 | 1810 | ||
| AK097433.1 | - | 12 | 1923 | |||
| BC136259.1 | - | 12 | 1877 | 9020246 | ||

ACRC
Genomic Map