siRNA Details
| Gene Symbol: SERHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027786.1 | 3 | 351 | |||
| GenBank mRNA | AK094872.1 | - | 3 | 351 | ||
| AL365513.1 | - | 3 | 340 | |||
| BC093888.1 | - | 3 | 305 | 7939733 | ||
| DA273263.1 | - | 3 | 351 | |||
| Gene Symbol: SERHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284334.1 | NP_001271263.1 | 2 | 268 | ||
| NM_014509.4 | NP_055324.2 | 3 | 418 | |||
| NR_104300.1 | 3 | 418 | ||||
| NR_104301.1 | 3 | 418 | ||||
| XM_017028738.1 | XP_016884227.1 | 3 | 407 | |||
| XM_017028739.1 | XP_016884228.1 | 3 | 405 | |||
| GenBank mRNA | AK294584.1 | - | 3 | 362 | ||
| AK295223.1 | - | 3 | 324 | |||
| AL450314.1 | - | 3 | 361 | |||
| AL590118.1 | - | 2 | 109 | |||
| AL590120.1 | - | 3 | 259 | |||
| BC119650.2 | - | 3 | 302 | 40115836 | ||
| BC119651.1 | - | 3 | 302 | 40115837 | ||
| BC127635.1 | - | 3 | 303 | 40115838 | ||
| BC127636.1 | - | 3 | 303 | 40115841 | ||
| BI603374.1 | - | 426 | ||||
| DC315058.1 | - | 3 | 395 | |||

SERHL
Genomic Map