siRNA Details
| Gene Symbol: TTYH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032646.5 | NP_116035.5 | 11, 12 | 1298 | ||
| NM_052869.1 | NP_443101.1 | 5, 6 | 1108 | |||
| XM_005257824.3 | XP_005257881.1 | 11, 12 | 1226 | |||
| GenBank mRNA | AF319952.1 | - | 11, 12 | 1277 | ||
| AK126955.1 | - | 5, 6 | 1516 | |||
| AK296511.1 | - | 11, 12 | 1221 | |||
| BC005168.1 | - | 11, 12 | 1273 | |||
| BC107602.1 | - | 11, 12 | 1294 | 40033791 | ||
| BX647912.1 | - | 5, 6 | 1108 | |||

TTYH2
Genomic Map