siRNA Details
| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 655 | ||
| NM_033204.3 | NP_149981.2 | 4 | 783 | |||
| GenBank mRNA | AL096747.1 | - | 3 | 2328 | ||
| AY149174.1 | - | 4 | 717 | |||
| BC011502.1 | - | 1 | 419 | |||
| BC094808.1 | - | 4 | 716 | |||
| BC119680.1 | - | 5 | 819 | 40116294 | ||

ZNF101
Genomic Map