siRNA Details
| Gene Symbol: DLG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166278.1 | NP_001159750.1 | 2 | 212 | ||
| NM_020730.2 | NP_065781.1 | 4 | 755 | |||
| NM_021120.3 | NP_066943.2 | 10 | 1796 | |||
| XM_005262248.3 | XP_005262305.1 | 4 | 352 | |||
| XM_006724625.2 | XP_006724688.1 | 10 | 1801 | |||
| XM_006724626.2 | XP_006724689.1 | 10 | 1801 | |||
| XM_011530883.1 | XP_011529185.1 | 10 | 1801 | |||
| XM_017029322.1 | XP_016884811.1 | 4 | 744 | |||
| XM_017029323.1 | XP_016884812.1 | 4 | 744 | |||
| XM_017029324.1 | XP_016884813.1 | 4 | 764 | |||
| XM_017029325.1 | XP_016884814.1 | 2 | 2062 | |||
| XM_017029326.1 | XP_016884815.1 | 2 | 141 | |||
| XM_017029327.1 | XP_016884816.1 | 2 | 207 | |||
| GenBank mRNA | AB033058.1 | - | 4 | 753 | ||
| AB209328.1 | - | 10 | 1796 | |||
| AK303377.1 | - | 2 | 212 | |||
| AK304020.1 | - | 4 | 468 | |||
| AK315041.1 | - | 10 | 1735 | |||
| AK316518.1 | - | 4 | 454 | |||
| BC093864.1 | - | 10 | 1497 | 7939709 | ||
| BC093866.1 | - | 10 | 1497 | 7939711 | ||
| U49089.1 | - | 10 | 1682 | |||

DLG3
Genomic Map