siRNA Details
| Gene Symbol: SYTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163334.1 | NP_001156806.1 | 12 | 1804 | ||
| NM_001163335.1 | NP_001156807.1 | 13 | 1916 | |||
| NM_138780.2 | NP_620135.1 | 12 | 1825 | |||
| XM_011544001.2 | XP_011542303.1 | 15 | 2243 | |||
| XM_011544002.2 | XP_011542304.1 | 13 | 1877 | |||
| XM_017029972.1 | XP_016885461.1 | 16 | 2409 | |||
| XM_017029973.1 | XP_016885462.1 | 15 | 2476 | |||
| XM_017029974.1 | XP_016885463.1 | 14 | 2104 | |||
| GenBank mRNA | AB080222.1 | - | 11 | 1382 | ||
| AK125106.1 | - | 12 | 1793 | |||
| BC131585.1 | - | 12 | 1448 | 40113205 | ||
| BX647688.1 | - | 13 | 1918 | |||

SYTL5
Genomic Map