siRNA Details
| Gene Symbol: FNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008738.2 | NP_001008738.2 | 10 | 1230 | ||
| NM_133372.2 | NP_588613.2 | 11 | 1314 | |||
| GenBank mRNA | AB075841.1 | - | 5 | 503 | ||
| AK292473.1 | - | 11 | 1268 | |||
| BC001956.1 | - | 11 | 1267 | |||
| CR749426.1 | - | 5 | 553 | |||
| CR936723.1 | - | 10 | 1067 | |||
| DQ145719.1 | - | 11 | 1219 | |||

FNIP1
Genomic Map