siRNA Details
| Gene Symbol: FNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008738.2 | NP_001008738.2 | 2, 3 | 354 | ||
| NM_133372.2 | NP_588613.2 | 2, 3 | 354 | |||
| GenBank mRNA | AK292473.1 | - | 2, 3 | 308 | ||
| BC001956.1 | - | 2, 3 | 307 | |||
| DQ145719.1 | - | 2, 3 | 259 | |||

FNIP1
Genomic Map