siRNA Details
| Gene Symbol: FNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008738.2 | NP_001008738.2 | 13 | 2476 | ||
| NM_133372.2 | NP_588613.2 | 14 | 2560 | |||
| GenBank mRNA | AB075841.1 | - | 8 | 1749 | ||
| AK292473.1 | - | 14 | 2514 | |||
| AL832008.1 | - | 1 | 572 | |||
| CR749426.1 | - | 9 | 1776 | |||
| CR936723.1 | - | 14 | 2290 | |||
| DQ145719.1 | - | 14 | 2465 | |||

FNIP1
Genomic Map