siRNA Details
| Gene Symbol: TBC1D27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025506.1 | XP_016880995.1 | 2 | 118 | ||
| GenBank mRNA | AK024458.1 | - | 11 | 2876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D27| Gene Symbol: TBC1D27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025506.1 | XP_016880995.1 | 2 | 118 | ||
| GenBank mRNA | AK024458.1 | - | 11 | 2876 | ||