siRNA Details
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244883.1 | NP_001231812.1 | 6, 7 | 386 | ||
NM_001933.4 | NP_001924.2 | 6, 7 | 386 | |||
NR_045209.1 | 5, 6 | 337 | ||||
GenBank mRNA | AK289414.1 | - | 6, 7 | 384 | ||
AK300396.1 | - | 6, 7 | 355 | |||
AU133381.1 | - | 6, 7 | 361 | |||
BC000302.2 | - | 6, 7 | 348 | 2820348 | ||
BC001922.1 | - | 6, 7 | 348 | 2820348 | ||
BX248774.1 | - | 5, 6 | 272 | |||
CR456727.1 | - | 6, 7 | 323 | |||
D16373.1 | - | 6, 7 | 335 | |||
L37418.1 | - | 6, 7 | 323 |
Gene Symbol: DLSTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130749.1 | 1 | 560 | |||
GenBank mRNA | S72422.1 | - | 1 | 558 |
