siRNA Details
| Gene Symbol: SAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320845.1 | NP_001307774.1 | 4 | 517 | ||
| NM_133491.4 | NP_597998.1 | 4 | 500 | |||
| XM_017024073.1 | XP_016879562.1 | 4 | 517 | |||
| GenBank mRNA | AF348524.1 | - | 4 | 259 | ||
| BC011751.2 | - | 4 | 470 | 3534187 | ||
| BC065922.1 | - | 1 | 809 | |||
| BC095466.1 | - | 4 | 455 | |||

SAT2
Genomic Map