siRNA Details
| Gene Symbol: TUBA3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207312.2 | NP_997195.1 | 4 | 1120 | ||
| GenBank mRNA | AA397983.1 | - | 1 | 397 | ||
| AK131043.1 | - | 4 | 1098 | |||
| BC057811.1 | - | 4 | 1120 | 5298532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBA3E| Gene Symbol: TUBA3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207312.2 | NP_997195.1 | 4 | 1120 | ||
| GenBank mRNA | AA397983.1 | - | 1 | 397 | ||
| AK131043.1 | - | 4 | 1098 | |||
| BC057811.1 | - | 4 | 1120 | 5298532 | ||