siRNA Details
| Gene Symbol: IL17F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052872.3 | NP_443104.1 | 3 | 403 | ||
| XM_011514276.1 | XP_011512578.1 | 4 | 469 | |||
| GenBank mRNA | AF332389.1 | - | 2 | 516 | ||
| AF384857.1 | - | 3 | 332 | |||
| BC070124.1 | - | 3 | 403 | 30417554 | ||
| JF305974.1 | - | 3 | 332 | |||

IL17F
Genomic Map