siRNA Details
Gene Symbol: IFT43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102564.1 | NP_001096034.1 | 9 | 569 | ||
NM_052873.2 | NP_443105.2 | 8 | 584 | |||
NR_045664.1 | 7 | 489 | ||||
NR_045665.1 | 9 | 569 | ||||
GenBank mRNA | AK056735.1 | - | 9 | 880 | ||
AK302944.1 | - | 9 | 552 | |||
BC010436.1 | - | 8 | 555 | 3608220 | ||
BI821833.1 | - | 7 | 465 | |||
CB853974.1 | - | 9 | 530 |
