siRNA Details
| Gene Symbol: PHF21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135862.2 | NP_001129334.1 | 14 | 2182 | ||
| NM_001242450.1 | NP_001229379.1 | 13 | 2222 | |||
| NM_001284296.1 | NP_001271225.1 | 14 | 2268 | |||
| NM_138415.4 | NP_612424.1 | 13 | 2353 | |||
| NR_134535.1 | 12 | 2114 | ||||
| XM_006724122.1 | XP_006724185.1 | 13 | 2362 | |||
| GenBank mRNA | AK091480.1 | - | 14 | 2268 | ||
| AK092243.1 | - | 14 | 2182 | |||
| AK095777.1 | - | 7 | 1334 | |||
| BC012187.1 | - | 13 | 2353 | 4634631 | ||

PHF21B
Genomic Map