siRNA Details
| Gene Symbol: CASC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138423.3 | NP_612432.2 | 5 | 954 | ||
| NM_177974.2 | NP_816929.1 | 5 | 954 | |||
| XM_017021880.1 | XP_016877369.1 | 5 | 1049 | |||
| GenBank mRNA | AK291026.1 | - | 5 | 711 | ||
| AK298564.1 | - | 6 | 669 | |||
| AL832333.1 | - | 5 | 726 | |||
| AL832840.1 | - | 5 | 396 | |||
| AY358086.1 | - | 5 | 905 | |||
| BC012124.1 | - | 4 | 843 | |||
| BC063480.1 | - | 5 | 908 | 5218924 | ||

CASC4
Genomic Map