siRNA Details
| Gene Symbol: SLC46A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242366.2 | NP_001229295.1 | 2 | 927 | ||
| NM_080669.5 | NP_542400.2 | 2 | 927 | |||
| XM_005277786.3 | XP_005277843.1 | 2 | 927 | |||
| XM_017024110.1 | XP_016879599.1 | 3 | 886 | |||
| GenBank mRNA | AK054669.1 | - | 2 | 925 | ||
| AK074161.1 | - | 1 | 25 | |||
| AK097194.1 | - | 1 | 68 | |||
| AK295883.1 | - | 2 | 883 | |||
| AL832613.2 | - | 1 | 235 | |||
| BC010691.1 | - | 2 | 916 | 3872267 | ||

SLC46A1
Genomic Map