siRNA Details
| Gene Symbol: GLCCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138426.3 | NP_612435.1 | 6 | 1774 | ||
| GenBank mRNA | AK126731.1 | - | 1 | 1046 | ||
| AL833694.1 | - | 2 | 269 | |||
| BC050291.1 | - | 6 | 1725 | 5272003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLCCI1| Gene Symbol: GLCCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138426.3 | NP_612435.1 | 6 | 1774 | ||
| GenBank mRNA | AK126731.1 | - | 1 | 1046 | ||
| AL833694.1 | - | 2 | 269 | |||
| BC050291.1 | - | 6 | 1725 | 5272003 | ||