siRNA Details
| Gene Symbol: GLCCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138426.3 | NP_612435.1 | 4 | 1333 | ||
| GenBank mRNA | BC050291.1 | - | 4 | 1284 | 5272003 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLCCI1| Gene Symbol: GLCCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138426.3 | NP_612435.1 | 4 | 1333 | ||
| GenBank mRNA | BC050291.1 | - | 4 | 1284 | 5272003 | |