siRNA Details
| Gene Symbol: DLX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005221.5 | NP_005212.1 | 2 | 639 | ||
| XM_005250185.3 | XP_005250242.1 | 2 | 1750 | |||
| XM_017011803.1 | XP_016867292.1 | 2 | 140 | |||
| GenBank mRNA | AK023493.1 | - | 2 | 639 | ||
| AK297614.1 | - | 2 | 759 | |||
| BC006226.2 | - | 2 | 625 | 3941691 | ||
| BT006903.1 | - | 2 | 431 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010056, AF022075, AF033011, AF072452, AK134920, and others.

DLX5
Genomic Map