siRNA Details
| Gene Symbol: HELQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297756.1 | NP_001284685.1 | 4 | 1519 | ||
| NM_001297757.1 | NP_001284686.1 | 4 | 1519 | |||
| NM_133636.3 | NP_598375.2 | 4 | 1519 | |||
| XM_005262711.1 | XP_005262768.1 | 4 | 1380 | |||
| XM_005262713.2 | XP_005262770.1 | 4 | 1609 | |||
| XM_006714076.2 | XP_006714139.1 | 4 | 1481 | |||
| XM_011531580.2 | XP_011529882.1 | 4 | 1481 | |||
| XM_017007681.1 | XP_016863170.1 | 4 | 1481 | |||
| XM_017007682.1 | XP_016863171.1 | 4 | 1587 | |||
| GenBank mRNA | AF436845.1 | - | 4 | 1480 | ||
| AK312725.1 | - | 4 | 1483 | |||
| AL512702.1 | - | 2 | 170 | |||
| BC111502.1 | - | 4 | 1439 | |||
| CK000270.1 | - | 1 | 119 | |||

HELQ
Genomic Map