siRNA Details
Gene Symbol: KLHDC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138433.3 | NP_612442.2 | 1 | 684 | ||
GenBank mRNA | BC009980.2 | - | 1 | 684 | 4121528 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLHDC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138433.3 | NP_612442.2 | 1 | 684 | ||
GenBank mRNA | BC009980.2 | - | 1 | 684 | 4121528 |