siRNA Details
| Gene Symbol: SLC2A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052885.3 | NP_443117.3 | 10 | 1875 | ||
| XM_011537847.2 | XP_011536149.1 | 9 | 1975 | |||
| XM_017018764.1 | XP_016874253.1 | 10 | 1348 | |||
| XM_017018765.1 | XP_016874254.1 | 10 | 1986 | |||
| XM_017018766.1 | XP_016874255.1 | 9 | 1133 | |||
| GenBank mRNA | AJ315644.1 | - | 10 | 1875 | ||
| BC117117.1 | - | 10 | 1819 | 40125668 | ||
| BC117119.1 | - | 10 | 1819 | 40125670 | ||

SLC2A13
Genomic Map