siRNA Details
| Gene Symbol: LACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288585.1 | NP_001275514.1 | 2, 3 | 496 | ||
| NM_032857.4 | NP_116246.2 | 2, 3 | 496 | |||
| NM_171846.3 | NP_741982.1 | 2, 3 | 496 | |||
| XM_011521180.1 | XP_011519482.1 | 2, 3 | 490 | |||
| GenBank mRNA | AK027808.1 | - | 2, 3 | 464 | ||
| AY358709.1 | - | 2, 3 | 444 | |||
| BC067288.1 | - | 2, 3 | 476 | 30366453 | ||

LACTB
Genomic Map