siRNA Details
| Gene Symbol: LACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288585.1 | NP_001275514.1 | 4 | 725 | ||
| NM_032857.4 | NP_116246.2 | 4 | 725 | |||
| NM_171846.3 | NP_741982.1 | 4 | 725 | |||
| XM_011521180.1 | XP_011519482.1 | 4 | 719 | |||
| GenBank mRNA | AK027808.1 | - | 4 | 693 | ||
| AY358709.1 | - | 4 | 673 | |||
| BC067288.1 | - | 4 | 705 | 30366453 | ||

LACTB
Genomic Map