siRNA Details
| Gene Symbol: NLRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079821.2 | NP_001073289.1 | 3 | 385 | ||
| NM_001127461.2 | NP_001120933.1 | 2 | 1027 | |||
| NM_001127462.2 | NP_001120934.1 | 1 | 993 | |||
| NM_001243133.1 | NP_001230062.1 | 1 | 993 | |||
| NM_004895.4 | NP_004886.3 | 1 | 993 | |||
| NM_183395.2 | NP_899632.1 | 1 | 993 | |||
| XM_011544048.2 | XP_011542350.1 | 2 | 1132 | |||
| XM_011544053.2 | XP_011542355.1 | 2 | 1020 | |||
| XM_011544055.2 | XP_011542357.1 | 2 | 1020 | |||
| XM_017000181.1 | XP_016855670.1 | 3 | 609 | |||
| XM_017000182.1 | XP_016855671.1 | 3 | 554 | |||
| XM_017000183.1 | XP_016855672.1 | 2 | 1020 | |||
| XM_017000184.1 | XP_016855673.1 | 2 | 1020 | |||
| GenBank mRNA | AB208891.1 | - | 1 | 1002 | ||
| AF410477.1 | - | 1 | 993 | |||
| AF418985.2 | - | 1 | 409 | |||
| AF420469.1 | - | 2 | 379 | |||
| AF427617.1 | - | 1 | 993 | |||
| AF468522.1 | - | 1 | 409 | |||
| AK027194.1 | - | 1 | 422 | |||
| AK314998.1 | - | 3 | 385 | |||
| AY092033.1 | - | 1 | 409 | |||
| AY422168.1 | - | 1 | 241 | |||
| BC117211.1 | - | 1 | 312 | 40125762 | ||
| BC143359.1 | - | 1 | 312 | 9051864 | ||
| BC143362.1 | - | 1 | 312 | 9051867 | ||
| BC143363.1 | - | 1 | 312 | 9051868 | ||
| DA244290.1 | - | 3 | 385 | |||

NLRP3
Genomic Map