siRNA Details
| Gene Symbol: CARD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017534.1 | NP_001017534.1 | 3 | 455 | ||
| NM_052889.2 | NP_443121.1 | 4 | 518 | |||
| XM_011542583.2 | XP_011540885.1 | 3 | 647 | |||
| GenBank mRNA | AY885669.1 | - | 3 | 446 | ||

CARD16
Genomic Map