siRNA Details
| Gene Symbol: MBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052897.3 | NP_443129.3 | 6 | 894 | ||
| XM_005268617.1 | XP_005268674.1 | 6 | 947 | |||
| XM_005268618.1 | XP_005268675.1 | 6 | 1043 | |||
| XM_005268620.2 | XP_005268677.1 | 6 | 947 | |||
| XM_006719217.1 | XP_006719280.1 | 6 | 792 | |||
| XM_006719218.1 | XP_006719281.1 | 6 | 780 | |||
| XM_011537851.2 | XP_011536153.1 | 6 | 1780 | |||
| GenBank mRNA | AB067474.1 | - | 3 | 563 | ||
| AK056399.1 | - | 6 | 928 | |||
| BC065530.1 | - | 6 | 894 | 6152748 | ||

MBD6
Genomic Map