siRNA Details
| Gene Symbol: KLHL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286250.1 | NP_001273179.1 | 6 | 1222 | ||
| NM_001286251.1 | NP_001273180.1 | 5 | 1123 | |||
| NM_001286252.1 | NP_001273181.1 | 7 | 1330 | |||
| NM_001323252.1 | NP_001310181.1 | 8 | 1377 | |||
| NM_001323253.1 | NP_001310182.1 | 7 | 1330 | |||
| NM_001323254.1 | NP_001310183.1 | 6 | 1114 | |||
| NM_001323255.1 | NP_001310184.1 | 6 | 1114 | |||
| NM_001323256.1 | NP_001310185.1 | 8 | 1409 | |||
| NM_001323257.1 | NP_001310186.1 | 6 | 1114 | |||
| NM_001323258.1 | NP_001310187.1 | 8 | 3070 | |||
| NM_001323259.1 | NP_001310188.1 | 7 | 1326 | |||
| NM_001323260.1 | NP_001310189.1 | 7 | 1326 | |||
| NM_001323261.1 | NP_001310190.1 | 5 | 840 | |||
| NM_001323262.1 | NP_001310191.1 | 5 | 840 | |||
| NM_001323263.1 | NP_001310192.1 | 5 | 844 | |||
| NM_001323264.1 | NP_001310193.1 | 4 | 628 | |||
| NM_052904.3 | NP_443136.2 | 7 | 1330 | |||
| XM_005266813.4 | XP_005266870.1 | 5 | 735 | |||
| XM_017010228.1 | XP_016865717.1 | 4 | 519 | |||
| XM_017010229.1 | XP_016865718.1 | 5 | 692 | |||
| GenBank mRNA | AB067487.1 | - | 5 | 744 | ||
| AK055292.1 | - | 7 | 1216 | |||
| AK295479.1 | - | 6 | 1047 | |||
| AK297240.1 | - | 5 | 867 | |||
| BC071627.1 | - | 7 | 1205 | 5245540 | ||
| BG181952.1 | - | 4 | 625 | |||
| DA255882.1 | - | 3 | 472 | |||

KLHL32
Genomic Map