siRNA Details
| Gene Symbol: DMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079911.2 | NP_001073380.1 | 5 | 900 | ||
| NM_004407.3 | NP_004398.1 | 6 | 948 | |||
| XM_011531705.2 | XP_011530007.1 | 6 | 1823 | |||
| XM_011531706.2 | XP_011530008.1 | 5 | 1775 | |||
| GenBank mRNA | BC130581.1 | - | 5 | 870 | 40146612 | |
| BC132865.1 | - | 5 | 870 | 40146887 | ||
| U65378.1 | - | 5 | 895 | |||
| U89012.1 | - | 6 | 943 | |||

DMP1
Genomic Map