siRNA Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052906.4 | NP_443138.2 | 3 | 1258 | ||
| GenBank mRNA | AB067491.1 | - | 1 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELFN2| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052906.4 | NP_443138.2 | 3 | 1258 | ||
| GenBank mRNA | AB067491.1 | - | 1 | 477 | ||