siRNA Details
| Gene Symbol: ESCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052911.2 | NP_443143.2 | 10 | 3104 | ||
| XM_011525798.1 | XP_011524100.1 | 10 | 3234 | |||
| XM_011525799.2 | XP_011524101.1 | 9 | 3277 | |||
| GenBank mRNA | AB067498.1 | - | 10 | 3040 | ||
| AK090579.1 | - | 7 | 862 | |||
| AL832041.1 | - | 11 | 3380 | |||
| AL834200.1 | - | 8 | 1936 | |||
| BC036943.1 | - | 4 | 473 | |||
| BC089426.1 | - | 7 | 2524 | 3090307 | ||

ESCO1
Genomic Map