siRNA Details
| Gene Symbol: RHPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052924.2 | NP_443156.2 | 6, 7 | 721 | ||
| XM_005250772.3 | XP_005250829.1 | 7, 8 | 907 | |||
| XM_005250773.3 | XP_005250830.1 | 7, 8 | 907 | |||
| XM_017013015.1 | XP_016868504.1 | 6, 7 | 1239 | |||
| XM_017013016.1 | XP_016868505.1 | 7, 8 | 907 | |||
| GenBank mRNA | AB067516.1 | - | 6, 7 | 579 | ||
| AY082588.1 | - | 6, 7 | 588 | |||
| BC025767.1 | - | 6, 7 | 689 | 5211354 | ||
| BX341691.2 | - | 6, 7 | 729 | |||

RHPN1
Genomic Map